Assay Details
| Gene Symbol: Cdkn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111099.1 | NP_001104569.1 | 2 - 3 | 556 | 80 | |
| NM_007669.4 | NP_031695.1 | 2 - 3 | 530 | 80 | ||
| GenBank mRNA | AB017817.1 | - | 1 - 2 | 429 | 80 | |
| AB017818.1 | - | 1 - 2 | 429 | 80 | ||
| AK007630.1 | - | 2 - 3 | 528 | 80 | ||
| AK089093.1 | - | 2 - 3 | 528 | 80 | ||
| AK139306.1 | - | 2 - 3 | 531 | 80 | ||
| AK149910.1 | - | 2 - 3 | 531 | 80 | ||
| AK170467.1 | - | 2 - 3 | 531 | 80 | ||
| BC002043.1 | - | 2 - 3 | 483 | 3495942 | 80 | |
| CT010218.1 | - | 1 - 2 | 429 | 80 | ||
| U09507.1 | - | 1 - 2 | 629 | 80 | ||
| U24173.1 | - | 2 - 3 | 521 | 80 | ||
| Gene Symbol: 4930567H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131763.1 | - | 812 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cdkn1a
Genomic Map