Assay Details
| Gene Symbol: Tmprss7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172455.3 | NP_766043.3 | 17 - 18 | 95 | ||
| GenBank mRNA | AK045663.1 | - | 15 - 16 | 95 | ||
| DQ061860.1 | - | 17 - 18 | 2324 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss7
Genomic Map