Assay Details
| Gene Symbol: S100a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163525.2 | NP_001156997.1 | 3 - 4 | 341 | 101 | |
| NM_025393.4 | NP_079669.2 | 3 - 4 | 324 | 101 | ||
| GenBank mRNA | AA726094.1 | - | 3 - 4 | 334 | 101 | |
| AK003669.1 | - | 3 - 4 | 304 | 101 | ||
| AK019075.1 | - | 3 - 4 | 295 | 101 | ||
| BC025607.1 | - | 2 - 3 | 323 | 5151924 | 101 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | AK132637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | BC025607.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | NM_001163526.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | NM_025393.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | NM_001163525.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | AK132637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | BC025607.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | NM_001163526.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | NM_025393.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419226 | NM_001163525.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281096 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180291 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | AK132637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | BC025607.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | NM_001163526.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | NM_025393.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | NM_001163525.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | AK132637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | BC025607.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | NM_001163526.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | NM_025393.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419226 | NM_001163525.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325747 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180291 | - | - |
| Whole Mouse Genome G4122A | A_51_P419226 | AK132637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | BC025607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | NM_001163526.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | NM_025393.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | NM_001163525.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | AK132637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | BC025607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | NM_001163526.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | NM_025393.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419226 | NM_001163525.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139241 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180291 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02741582 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02741578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02741598 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02741607 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02741602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02741579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02424826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493796 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449166_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493794 | - | - |
| Mouse Genome 430 2.0 Array | 1449166_at | - | - |
| Mouse Genome 430 2.0 Array | 1449166_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66166_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66166_PM_at | - | - |
| Mouse Expression Set 430 | 1449166_PM_at | - | - |
| Mouse Expression Set 430 | 1449166_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449166_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449166_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449166_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449166_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 450684 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 450400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 450400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000450400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777308 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2777308 | - | - |
| mouse-6 v1.1 (Illumina) | scl22946.3.1_72-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 450400 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22946.3.1_72-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 450400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2779858 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2779858 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2777308 | - | - |

S100a14
Genomic Map