Assay Details
| Gene Symbol: Hnrnpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170981.1 | NP_001164452.1 | 2 - 3 | 167 | 102 | |
| NM_001170983.1 | NP_001164454.1 | 2 - 3 | 167 | 102 | ||
| NM_001170984.1 | NP_001164455.1 | 2 - 3 | 167 | 102 | ||
| NM_016884.3 | NP_058580.1 | 2 - 3 | 167 | 102 | ||
| GenBank mRNA | AK010845.1 | - | 2 - 3 | 146 | 102 | |
| AK011336.1 | - | 2 - 3 | 139 | 102 | ||
| AK012633.2 | - | 2 - 3 | 129 | 102 | ||
| AK088245.1 | - | 2 - 3 | 144 | 102 | ||
| AK088678.1 | - | 2 - 3 | 139 | 102 | ||
| AK089061.1 | - | 2 - 3 | 147 | 103 | ||
| AK151446.1 | - | 2 - 3 | 144 | 102 | ||
| AK153053.1 | - | 2 - 3 | 144 | 102 | ||
| AK166590.1 | - | 2 - 3 | 145 | 102 | ||
| AK168802.1 | - | 2 - 3 | 141 | 102 | ||
| AK169287.1 | - | 2 - 3 | 139 | 102 | ||
| BC004706.1 | - | 2 - 3 | 129 | 3499283 | 102 | |
| DV652725.1 | - | 2 - 3 | 191 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011336.1,AK012633.2,AK088245.1,AK088678.1,AK089061.1,AK153053.1,AK166590.1,AK168802.1,BC004706.1,NM_001170981.1,NM_001170983.1,NM_001170984.1,NM_016884.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpc
Genomic Map