Assay Details
| Gene Symbol: Hic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010430.2 | NP_034560.2 | 1 - 2 | 1255 | 107 | |
| GenBank mRNA | AJ585343.1 | - | 1 - 2 | 1252 | 107 | |
| CJ067923.1 | - | 1 - 2 | 145 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hic1
Genomic Map