Assay Details
| Gene Symbol: Pira2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011089.2 | NP_035219.2 | 8 - 9 | 1882 | 75 | |
| GenBank mRNA | AK140776.1 | - | 8 - 9 | 1888 | 75 | |
| AK172046.1 | - | 1883 | 75 | |||
| U96683.1 | - | 1849 | 75 | |||
| Gene Symbol: Pira1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011087.1 | NP_035217.1 | 1863 | 75 | ||
| GenBank mRNA | U96682.1 | - | 1863 | 75 | ||
| Gene Symbol: Pira11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011088.1 | NP_035218.1 | 1863 | 75 | ||
| GenBank mRNA | BC103528.2 | - | 1863 | 40039374 | 75 | |
| BC103529.1 | - | 1863 | 40039371 | 75 | ||
| BC139803.1 | - | 1578 | 40039373 | 75 | ||
| BC150155.1 | - | 1581 | 40141323 | 75 | ||
| BC150712.1 | - | 1610 | 9087623 | 75 | ||
| Gene Symbol: Pira4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011091.1 | NP_035221.1 | 1862 | 75 | ||
| GenBank mRNA | U96685.1 | - | 1862 | 75 | ||
| Gene Symbol: Pira5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U96686.1 | - | 8 - 9 | 1566 | 75 | |
| Gene Symbol: Pira7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011094.1 | NP_035224.1 | 9 - 10 | 1890 | 75 | |
| GenBank mRNA | U96688.1 | - | 9 - 10 | 1890 | 75 | |
| Gene Symbol: Pirb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK165650.1 | - | 2039 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pira2
Genomic Map