Assay Details
| Gene Symbol: Mxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008542.2 | NP_001008542.2 | 3 - 4 | 631 | 96 | |
| NM_001008543.2 | NP_001008543.1 | 3 - 4 | 318 | 96 | ||
| NM_010847.3 | NP_034977.1 | 3 - 4 | 471 | 96 | ||
| GenBank mRNA | AK140465.1 | - | 3 - 4 | 421 | 96 | |
| AK144219.1 | - | 3 - 4 | 513 | 96 | ||
| AK154538.1 | - | 3 - 4 | 621 | 96 | ||
| BC064453.1 | - | 3 - 4 | 616 | 6810025 | 96 | |
| D31824.1 | - | 3 - 4 | 471 | 96 | ||
| L38821.1 | - | 3 - 4 | 324 | 96 | ||
| L38822.1 | - | 3 - 4 | 343 | 96 | ||
| U24673.1 | - | 3 - 4 | 304 | 96 | ||
| U24674.1 | - | 3 - 4 | 253 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mxi1
Genomic Map