| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700011860.mm.1 |
NM_001277980.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011860.mm.1 |
NM_030104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011860.mm.1 |
NM_030104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011860.mm.1 |
TC1700001301.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700011863.mm.1 |
NM_001277980.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011863.mm.1 |
NM_030104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011863.mm.1 |
NM_030104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011863.mm.1 |
TC1700001301.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700011861.mm.1 |
NM_001277980.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011861.mm.1 |
NM_030104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011861.mm.1 |
NM_030104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700011861.mm.1 |
TC1700001301.mm.1 |
MTA TC |