Assay Details
| Gene Symbol: Snapin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133854.3 | NP_598615.1 | 3 - 4 | 348 | 113 | |
| GenBank mRNA | AF086838.1 | - | 3 - 4 | 309 | 113 | |
| AK007458.1 | - | 3 - 4 | 329 | 113 | ||
| AK035147.1 | - | 3 - 4 | 345 | 113 | ||
| AK044085.1 | - | 2 - 3 | 282 | 113 | ||
| AK048007.1 | - | 3 - 4 | 337 | 113 | ||
| AK050151.1 | - | 3 - 4 | 349 | 113 | ||
| AK077840.1 | - | 3 - 4 | 339 | 113 | ||
| AK080718.1 | - | 3 - 4 | 339 | 113 | ||
| AK086994.1 | - | 2 - 3 | 280 | 113 | ||
| AK135006.1 | - | 3 - 4 | 340 | 113 | ||
| AK152057.1 | - | 3 - 4 | 317 | 113 | ||
| BC006744.1 | - | 1 - 2 | 275 | 3708620 | 113 | |
| BC048691.1 | - | 3 - 4 | 320 | 6511395 | 113 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snapin
Genomic Map