Assay Details
| Gene Symbol: H1foo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138311.2 | NP_612184.1 | 3 - 4 | 708 | 92 | |
| GenBank mRNA | AK135944.1 | - | 3 - 4 | 712 | 92 | |
| AK162137.1 | - | 3 - 4 | 687 | 92 | ||
| AK162154.1 | - | 3 - 4 | 705 | 92 | ||
| AK165127.1 | - | 4 - 5 | 790 | 92 | ||
| AY007195.1 | - | 3 - 4 | 708 | 92 | ||
| BC137916.1 | - | 3 - 4 | 704 | 8860935 | 92 | |
| BC145046.1 | - | 3 - 4 | 704 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | AK162137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | AK162154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | AK165127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | AY007195.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | NM_138311.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | AK162137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | AK162154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | AK165127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | AY007195.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P178815 | NM_138311.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909240 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659452 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129934 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | AK162137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | AK162154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | AK165127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | AY007195.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | NM_138311.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | AK162137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | AK162154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | AK165127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | AY007195.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P178815 | NM_138311.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | AK162137.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | AK162154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | AK165127.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | AY007195.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | NM_138311.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | AK162137.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | AK162154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | AK165127.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | AY007195.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178815 | NM_138311.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24773918 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129934 | - | - |
| Whole Mouse Genome G4122A | A_51_P178815 | AK162137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | AK162154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | AK165127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | AY007195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | NM_138311.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | AK162137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | AK162154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | AK165127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | AY007195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P178815 | NM_138311.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108927 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129934 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05052005 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05052014 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05052009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05809322 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05052013 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05052010 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05052006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24773958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05294179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05294183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05052016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05352416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05700316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05809321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05783781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05750576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05682936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05779620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05809322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05779621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05682935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05052014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05356142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05310728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05052009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05682937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05008235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05874512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05809323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05750575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05534139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05847842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05682934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05336980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05052006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05336981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24773916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05310722 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10540996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540997 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540998 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540994 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416518_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10540993 | - | - |
| Mouse Genome 430 2.0 Array | 1416518_at | - | - |
| Mouse Genome 430 2.0 Array | 1416518_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171506_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171506_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600010237.mm.1 | NM_138311.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600010237.mm.1 | TC0600001290.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600010240.mm.1 | NM_138311.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600010240.mm.1 | TC0600001290.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600010239.mm.1 | NM_138311.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600010239.mm.1 | TC0600001290.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416518_PM_at | - | - |
| Mouse Expression Set 430 | 1416518_at | - | - |
| Murine Genome U74v2 Set | 160147_PM_r_at | - | - |
| Murine Genome U74v2 Set | 160147_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416518_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416518_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416518_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416518_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 561765 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5700041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5700041 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2749809 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2749809 | - | - |
| mouse-6 v1.1 (Illumina) | scl29601.6.1_119-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700041 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29601.6.1_119-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700041 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2749809 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2749809 | - | - |

H1foo
Genomic Map