| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700004683.mm.1 |
NM_011981.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004683.mm.1 |
NM_011981.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004683.mm.1 |
TC0700000546.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004684.mm.1 |
NM_011981.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004684.mm.1 |
NM_011981.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004684.mm.1 |
TC0700000546.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004672.mm.1 |
NM_011981.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004672.mm.1 |
NM_011981.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004672.mm.1 |
TC0700000546.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004677.mm.1 |
NM_011981.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004677.mm.1 |
NM_011981.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004677.mm.1 |
TC0700000546.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004681.mm.1 |
NM_011981.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004681.mm.1 |
NM_011981.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004681.mm.1 |
TC0700000546.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004668.mm.1 |
NM_011981.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004668.mm.1 |
NM_011981.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004668.mm.1 |
TC0700000546.mm.1 |
MTA TC |