Assay Details
| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153106.2 | NP_694746.2 | 12 - 13 | 1522 | 90 | |
| GenBank mRNA | AF529423.1 | - | 12 - 13 | 1498 | 90 | |
| AK139616.1 | - | 12 - 13 | 1523 | 90 | ||
| BC053724.1 | - | 12 - 13 | 1520 | 30051727 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Padi6
Genomic Map