Assay Details
| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153106.2 | NP_694746.2 | 15 - 16 | 1885 | 89 | |
| GenBank mRNA | AF529423.1 | - | 15 - 16 | 1861 | 89 | |
| AK139616.1 | - | 15 - 16 | 1886 | 89 | ||
| BC053724.1 | - | 15 - 16 | 1883 | 30051727 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044498 | CJ202890.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044498 | NM_153106.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924983 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125765 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124784 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169496 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044498 | CJ202890.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044498 | NM_153106.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P448094 | BC053724.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P448094 | CJ202890.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P448094 | AF529423.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P448094 | AK139616.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P448094 | NM_153106.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23359945 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01006167 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05428502 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169496 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124784 | - | - |
| Whole Mouse Genome G4122A | A_51_P448094 | BC053724.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P448094 | CJ202890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P448094 | AF529423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P448094 | AK139616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P448094 | NM_153106.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103296 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169496 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124784 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03881188 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03452386 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03730147 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04032676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23359964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03746320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03633022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03593067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03452389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03683421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03738585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03411736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03730147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03927114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03683403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04019128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04032676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03378090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03613283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03881189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03944422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03944423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03790300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23359947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03927115 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517785 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517779 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517786 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517775 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517788 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517782 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517787 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517784 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517789 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517780 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437084_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517774 | - | - |
| Mouse Genome 430 2.0 Array | 1437084_at | - | - |
| Mouse Genome 430 2.0 Array | 1437084_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242726_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242726_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400031857.mm.1 | NM_153106.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031857.mm.1 | NM_153106.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031857.mm.1 | TC0400003864.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437084_at | - | - |
| Mouse Expression Set 430 | 1437084_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437084_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437084_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5080441 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5080441 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005080441 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216003 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216003 | - | - |
| mouse-6 v1.1 (Illumina) | scl23609.18.1_108-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5080441 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23609.18.1_108-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080441 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1216003 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216003 | - | - |

Padi6
Genomic Map