Assay Details
| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153106.2 | NP_694746.2 | 5 - 6 | 586 | 81 | |
| GenBank mRNA | AK139616.1 | - | 5 - 6 | 587 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Padi6
Genomic Map