Assay Details
| Gene Symbol: Snhg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175692.3 | NP_783623.2 | 4 - 5 | 661 | 102 | |
| GenBank mRNA | AK080745.1 | - | 4 - 5 | 619 | 102 | |
| AK157790.1 | - | 5 - 6 | 718 | 102 | ||
| BC030414.1 | - | 4 - 5 | 577 | 5366924 | 102 | |
| BC049241.1 | - | 575 | 4505955 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21782585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21782587 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21782586 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047809 | AK157790.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047809 | BC030414.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047809 | BC049241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047809 | AK080745.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047809 | NM_175692.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267933 | BC058997.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267933 | AK149064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267933 | BC028327.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267933 | NM_175692.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267933 | BC058997.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267933 | AK149064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267933 | BC028327.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267933 | NM_175692.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716385 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 138875 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163098 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115361 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267933 | BC058997.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267933 | AK149064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267933 | BC028327.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267933 | NM_175692.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267933 | BC058997.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267933 | AK149064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267933 | BC028327.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267933 | NM_175692.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047809 | AK157790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047809 | BC030414.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047809 | BC049241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047809 | AK080745.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047809 | NM_175692.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21782574 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00538811 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115361 | - | - |
| Whole Mouse Genome G4122A | A_52_P70888 | BC058997.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P70888 | AK149064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P70888 | BC028327.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P70888 | NM_175692.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385114 | AK157790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385114 | BC030414.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385114 | BC049241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385114 | AK080745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385114 | NM_175692.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541118 | AK157790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541118 | BC030414.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541118 | BC049241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541118 | AK080745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541118 | NM_175692.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267933 | BC058997.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267933 | AK149064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267933 | BC028327.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267933 | NM_175692.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267933 | BC058997.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267933 | AK149064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267933 | BC028327.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267933 | NM_175692.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108668 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102821 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115361 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02095165 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02095168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01869204 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02178706 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02095153 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21782585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21782586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21782587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21782602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01933266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01960785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01366900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01380780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01612311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02024900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01867275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02096398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01612309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02014398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02095162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02226679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02014399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01852944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02024903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02096396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02196529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01612310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01787416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01949138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02112360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02178706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02014401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02024897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02096395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01917072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21782666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02227292 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10478074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478071 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434292_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437311_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478073 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478066 | - | - |
| Mouse Genome 430 2.0 Array | 1437311_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437311_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319317_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319317_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200019716.mm.1 | NM_175692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019716.mm.1 | NM_175692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019716.mm.1 | TC0200002427.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019710.mm.1 | NM_175692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019710.mm.1 | NM_175692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019710.mm.1 | TC0200002427.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019712.mm.1 | NM_175692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019712.mm.1 | NM_175692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019712.mm.1 | TC0200002427.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019714.mm.1 | NM_175692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019714.mm.1 | NM_175692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019714.mm.1 | TC0200002427.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019711.mm.1 | NM_175692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019711.mm.1 | NM_175692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019711.mm.1 | TC0200002427.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019715.mm.1 | NM_175692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019715.mm.1 | NM_175692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019715.mm.1 | TC0200002427.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019717.mm.1 | NM_175692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019717.mm.1 | NM_175692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019717.mm.1 | TC0200002427.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437311_at | - | - |
| Mouse Expression Set 430 | 1437311_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434292_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434292_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437311_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437311_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 648531 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4010332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004010332 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2644381 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238627 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2644381 | - | - |
| mouse-6 v1.1 (Illumina) | scl20001.6.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4010332 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20001.6.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4010332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2952098 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2952098 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20001.6.1_12-S | - | - |

Snhg11
Genomic Map