Assay Details
| Gene Symbol: Ispd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289502.1 | NP_001276431.1 | 4 - 5 | 1000 | 108 | |
| NM_001289503.1 | NP_001276432.1 | 5 - 6 | 732 | 108 | ||
| NM_178629.6 | NP_848744.2 | 5 - 6 | 1150 | 108 | ||
| GenBank mRNA | AK016317.1 | - | 5 - 6 | 699 | 108 | |
| AK039004.1 | - | 5 - 6 | 1018 | 108 | ||
| AK045940.1 | - | 5 - 6 | 1152 | 108 | ||
| AK051322.1 | - | 4 - 5 | 695 | 108 | ||
| BC024061.1 | - | 4 - 5 | 432 | 108 | ||
| BC050993.1 | - | 5 - 6 | 988 | 108 | ||
| BC086665.1 | - | 5 - 6 | 1081 | 30612014 | 108 | |
| BC118030.1 | - | 1 - 2 | 108 | |||
| BC118038.1 | - | 5 - 6 | 918 | 40093984 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ispd
Genomic Map