Assay Details
| Gene Symbol: Set | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023871.4 | NP_076360.1 | 1 | 95 | 67 | |
| GenBank mRNA | AK014291.1 | - | 1 | 213 | 67 | |
| AK031561.1 | - | 1 | 211 | 67 | ||
| AK054400.1 | - | 1 | 68 | |||
| AK172327.1 | - | 1 | 239 | 67 | ||
| BC018255.1 | - | 1 | 48 | 4014298 | 67 | |
| BE331951.1 | - | 1 | 95 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014291.1, AK172327.1, BC018255.1, NM_023871.4 transcripts.
Set Membership: |
|

Set
Genomic Map