Assay Details
| Gene Symbol: Ndufc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024220.2 | NP_077182.1 | 1 | 49 | 87 | |
| GenBank mRNA | CK128102.1 | - | 1 | 49 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_024220.2 transcripts.
Set Membership: |
|

Ndufc2
Genomic Map