Assay Details
| Gene Symbol: Romo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025946.6 | NP_080222.1 | 1 | 46 | 71 | |
| GenBank mRNA | EL963857.1 | - | 1 | 56 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_025946.6 transcripts.
Set Membership: |
|

Romo1
Genomic Map