Assay Details
| Gene Symbol: Mir99b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029536.1 | 1 | 38 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
vomeronasal 2, receptor 90 |
Vmn2r90 | EG626942 | -96.0kb | Sense | Chr. 17 - 17669137 - 17734167 on NCBI Build GRCm38 |
| Downstream | microRNA let7e | Mirlet7e |
Mirnlet7e, mmu-let-7e |
95.0 | Sense | Chr. 17 - 17830352 - 17830444 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir99b
Genomic Map