Assay Details
| Gene Symbol: AI414108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027907.1 | 1 | 3743 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
immunoglobulin superfamily, member 9B |
Igsf9b |
Gm508, mKIAA1030 |
Sense | Chr. 9 - 27299104 - 27357549 on NCBI Build GRCm38 | |
| Downstream | spermatogenesis associated 19 | Spata19 |
1700001L23Rik, AV047838, spergen-1 |
39.3kb | Sense | Chr. 9 - 27396821 - 27401711 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AI414108
Genomic Map