Assay Details
| Gene Symbol: Gm4566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028023.1 | 2 - 3 | 228 | 79 | ||
| GenBank mRNA | CO802078.1 | - | 2 - 3 | 230 | 79 | |
| DV658461.1 | - | 1 - 2 | 350 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | elastin microfibril interfacer 2 | Emilin2 | FOAP-10 | -21.6kb | Sense | Chr. 17 - 71252172 - 71311639 on NCBI Build GRCm38 |
| Downstream | SMC hinge domain containing 1 | Smchd1 |
4931400A14Rik, AW554188, MommeD1, mKIAA0650 |
2.9kb | Sense | Chr. 17 - 71344489 - 71475343 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm4566
Genomic Map