Assay Details
| Gene Symbol: Hk1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038038.1 | 1 - 2 | 171 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tachykinin receptor 2 | Tacr2 |
Nk2r, Skr, Tac2r |
-74.6kb | Sense | Chr. 10 - 62252438 - 62265990 on NCBI Build GRCm38 |
| Downstream | hexokinase domain containing 1 | Hkdc1 | BC016235 | 33.3kb | Anti-sense | Chr. 10 - 62383137 - 62422465 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Hk1os
Genomic Map