Assay Details
Gene Symbol: Gm12238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028480.1 | 1 | 62 | 63 | ||
GenBank mRNA | AF357392.1 | - | 1 | 62 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GTPase activating protein (SH3 domain) binding protein 1 | G3bp1 |
AI849976, B430204O07, C87777, G3bp, mKIAA4115 |
Sense | Chr. 11 - 55469752 - 55500888 on NCBI Build GRCm38 | |
Downstream | microRNA 7652 | Mir7652 |
Gm27932, mmu-mir-7652 |
18.4kb | Sense | Chr. 11 - 55501226 - 55501281 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
