Assay Details
| Gene Symbol: Mir128-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029543.1 | 1 | 32 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | R3H domain containing 1 | R3hdm1 | R3hdm | Sense | Chr. 1 - 128103293 - 128237738 on NCBI Build GRCm38 | |
| Downstream | UBX domain protein 4 | Ubxn4 |
1300013G12Rik, Ubxd2, mKIAA0242 |
41.5kb | Sense | Chr. 1 - 128243898 - 128279378 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir128-1
Genomic Map