Assay Details
| Gene Symbol: Mirlet7e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029730.1 | 1 | 45 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 99b | Mir99b |
Mirn99b, mmu-mir-99b |
-95.0 | Sense | Chr. 17 - 17830188 - 17830257 on NCBI Build GRCm38 |
| Downstream | microRNA 125a | Mir125a | Mirn125a | 368.0 | Sense | Chr. 17 - 17830812 - 17830879 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7e
Genomic Map