| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
62201 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
267027 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58028 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
76795 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45633 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
698555 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
791687 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
896121 |
XM_904376.3 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
57160 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676962 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758834 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
382086 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
39866 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
110223 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510450 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351433 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497636 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
279910 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933181 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
59794 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
22983 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
29228 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118234 |
NR_033501.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118234 |
BC147734.1 |
mouse_transcriptome |