Assay Details
| Gene Symbol: Olfr1077-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033507.1 | 1 - 2 | 47 | 106 | ||
| GenBank mRNA | DR065539.1 | - | 1 - 2 | 47 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | olfactory receptor 1076 | Olfr1076 | MOR189-2 | -16.3kb | Anti-sense | Chr. 2 - 86508461 - 86509402 on NCBI Build GRCm38 |
| Downstream | olfactory receptor 1079 | Olfr1079 | MOR189-1 | 9.5kb | Sense | Chr. 2 - 86537966 - 86542480 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Olfr1077-ps1
Genomic Map