Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_001177601.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_001177601.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_001177603.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_001177603.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_001177606.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_001177606.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_001177607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_001177607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_026757.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_026757.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_145758.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
NM_145758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028476.mm.1 |
TC1100003089.mm.1 |
MTA TC |