Assay Details
| Gene Symbol: S100a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195760.1 | NP_001182689.1 | 1 - 2 | 107 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

S100a2| Gene Symbol: S100a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195760.1 | NP_001182689.1 | 1 - 2 | 107 | 91 | |
Set Membership: |
|