Assay Details
| Gene Symbol: Set | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204875.1 | NP_001191804.1 | 1 - 2 | 419 | 96 | |
| GenBank mRNA | AB015613.1 | - | 1 - 2 | 83 | 96 | |
| AK019960.1 | - | 1 - 2 | 168 | 96 | ||
| BQ899734.1 | - | 1 - 2 | 419 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Set
Genomic Map