| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200004030.mm.1 |
NM_001204875.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004030.mm.1 |
NM_001204875.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004030.mm.1 |
NM_023871.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004030.mm.1 |
NM_023871.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004030.mm.1 |
TC0200000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200004032.mm.1 |
NM_001204875.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004032.mm.1 |
NM_001204875.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004032.mm.1 |
NM_023871.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004032.mm.1 |
NM_023871.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004032.mm.1 |
TC0200000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200004029.mm.1 |
NM_001204875.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004029.mm.1 |
NM_001204875.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004029.mm.1 |
NM_023871.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004029.mm.1 |
NM_023871.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004029.mm.1 |
TC0200000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200004031.mm.1 |
NM_001204875.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004031.mm.1 |
NM_001204875.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004031.mm.1 |
NM_023871.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004031.mm.1 |
NM_023871.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004031.mm.1 |
TC0200000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200004033.mm.1 |
NM_001204875.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004033.mm.1 |
NM_001204875.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004033.mm.1 |
NM_023871.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004033.mm.1 |
NM_023871.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004033.mm.1 |
TC0200000522.mm.1 |
MTA TC |