Assay Details
| Gene Symbol: Klk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174866.2 | NP_777355.1 | 4 - 5 | 604 | 92 | |
| GenBank mRNA | BC044756.2 | - | 3 - 4 | 579 | 92 | |
| BC128019.1 | - | 4 - 5 | 607 | 40136266 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
fertilization
negative regulation of G-protein coupled receptor protein signaling pathway
positive regulation of G-protein coupled receptor protein signaling pathway
epidermis morphogenesis
seminal clot liquefaction
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity
proteolysis
fertilization
negative regulation of G-protein coupled receptor protein signaling pathway
positive regulation of G-protein coupled receptor protein signaling pathway
epidermis morphogenesis
seminal clot liquefaction
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084950 | NM_174866.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084950 | BC044756.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084950 | BC128019.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79150 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122311 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084950 | NM_174866.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084950 | BC044756.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084950 | BC128019.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083671 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122311 | - | - |
| Whole Mouse Genome G4122A | A_52_P294365 | NM_174866.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P294365 | BC044756.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P294365 | BC128019.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06081855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06364531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05982015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06148182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05981990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06364530 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552467 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552463 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552462 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700006246.mm.1 | NM_174866.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006246.mm.1 | TC0700000769.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006245.mm.1 | NM_174866.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006245.mm.1 | TC0700000769.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 763017 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4850605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850605 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2613722 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2613722 | - | - |
| mouse-6 v1.1 (Illumina) | scl32746.4.1_63-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850605 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32746.4.1_63-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2613722 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2613722 | - | - |

Klk14
Genomic Map