| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300001961.mm.1 |
NR_045075.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001961.mm.1 |
NR_045075.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001961.mm.1 |
NR_045077.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001961.mm.1 |
NR_045077.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001961.mm.1 |
TC1300000317.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001945.mm.1 |
NR_045077.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001945.mm.1 |
NR_045077.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001945.mm.1 |
TC1300000317.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001956.mm.1 |
NR_045074.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001956.mm.1 |
NR_045074.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001956.mm.1 |
NR_045077.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001956.mm.1 |
NR_045077.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001956.mm.1 |
TC1300000317.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001944.mm.1 |
NR_045077.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001944.mm.1 |
NR_045077.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001944.mm.1 |
TC1300000317.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001955.mm.1 |
NR_045074.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001955.mm.1 |
NR_045074.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001955.mm.1 |
NR_045077.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001955.mm.1 |
NR_045077.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001955.mm.1 |
TC1300000317.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001958.mm.1 |
NR_045073.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001958.mm.1 |
NR_045073.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001958.mm.1 |
NR_045075.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001958.mm.1 |
NR_045075.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001958.mm.1 |
NR_045077.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001958.mm.1 |
NR_045077.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001958.mm.1 |
TC1300000317.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001960.mm.1 |
NR_045075.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001960.mm.1 |
NR_045075.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001960.mm.1 |
NR_045077.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001960.mm.1 |
NR_045077.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001960.mm.1 |
TC1300000317.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
NR_045073.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
NR_045073.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
NR_045074.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
NR_045074.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
NR_045075.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
NR_045075.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
NR_045077.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
NR_045077.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001954.mm.1 |
TC1300000317.mm.1 |
MTA TC |