Assay Details
Gene Symbol: 1110015O18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045272.1 | 2 - 3 | 204 | 139 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
hepatocyte nuclear factor 4, gamma |
Hnf4g | NR2A2 | -1.1mb | Sense | Chr. 3 - 3508030 - 3659800 on NCBI Build GRCm38 |
Downstream |
predicted gene, 17308 |
Gm17308 | 289.5kb | Sense | Chr. 3 - 5090276 - 5093402 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
