| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
381229 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
727856 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386793 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
72525 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758408 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
801470 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
247257 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
215035 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
465322 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
47539 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
78594 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
243268 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
122355 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
312042 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
593685 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
641965 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
55781 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
525672 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
295505 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
492752 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
470307 |
L19714.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
744533 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
915645 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776607 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
395193 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
792110 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917388 |
AF449425.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917388 |
AK030396.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917388 |
AK043757.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917388 |
AK122190.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917388 |
BC024719.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917388 |
NM_133241.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
62080 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
146953 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
120702 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
35877 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
43848 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P131889 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05570478 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P131889 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05570544 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05570470 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05570503 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P312473 |
AK136375.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P312473 |
AK163067.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P312473 |
NR_015537.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P312473 |
AK050940.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125146 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05570503 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P131889 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693639 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693407 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04941187 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693636 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693405 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693632 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04710183 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693626 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693630 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693634 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04941190 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693637 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04941186 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04710185 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693412 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23693628 |
- |
- |