Assay Details
| Gene Symbol: Rnf223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001220499.1 | NP_001207428.1 | 1 | 213 | 66 | |
| GenBank mRNA | AK137636.1 | - | 1 | 1870 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf223
Genomic Map