Assay Details
| Gene Symbol: Fth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073181.1 | 1 - 2 | 287 | 130 | ||
| GenBank mRNA | AK152702.1 | - | 1 - 2 | 289 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK152702.1 transcripts.
Set Membership: |
|

Fth1
Genomic Map