Assay Details
| Gene Symbol: Bin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270537.1 | NP_001257466.1 | 4 - 5 | 315 | 78 | |
| GenBank mRNA | BY751384.1 | - | 4 - 5 | 318 | 78 | |
| CA562068.1 | - | 4 - 5 | 304 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bin2
Genomic Map