Assay Details
| Gene Symbol: Gm5741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195531.1 | NP_001182460.1 | 1 - 2 | 176 | 90 | |
| GenBank mRNA | BC147108.1 | - | 1 - 2 | 176 | 9056151 | 90 |
| BC147109.1 | - | 1 - 2 | 176 | 9056141 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gm5741
Genomic Map