Assay Details
| Gene Symbol: Rasd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009026.4 | NP_033052.1 | 1 - 2 | 423 | 61 | |
| GenBank mRNA | AF009246.1 | - | 1 - 2 | 427 | 61 | |
| AK038932.1 | - | 1 - 2 | 426 | 61 | ||
| BC034166.1 | - | 1 - 2 | 393 | 4989312 | 61 | |
| CT010376.1 | - | 1 - 2 | 286 | 61 | ||
More Information
Set Membership: |
|

Rasd1
Genomic Map