Assay Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104446.1 | 1 - 2 | 84 | 68 | ||
| GenBank mRNA | BC027091.1 | - | 1 - 2 | 77 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC027091.1 transcripts.
Set Membership: |
|

Rtn1
Genomic Map