Assay Details
| Gene Symbol: Mir6997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105963.1 | 1 | 42 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
spectrin alpha, non-erythrocytic 1 |
Sptan1 |
2610027H02Rik, Spna-2, Spna2 |
Sense | Chr. 2 - 29965553 - 30031451 on NCBI Build GRCm38 | |
| Downstream | WD repeat domain 34 | Wdr34 | 3200002I06Rik | 35.5kb | Anti-sense | Chr. 2 - 30031551 - 30048879 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6997
Genomic Map