Assay Details
| Gene Symbol: Mir7240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106099.1 | 1 | 29 | 50 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microtubule associated serine/threonine kinase 3 | Mast3 |
BC024265, mKIAA0561 |
Anti-sense | Chr. 8 - 70778117 - 70805068 on NCBI Build GRCm38 | |
| Downstream |
interleukin 12 receptor, beta 1 |
Il12rb1 |
CD212, IL-12R[b] |
10.2kb | Sense | Chr. 8 - 70808360 - 70821424 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7240
Genomic Map