Assay Details
| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110963.1 | 3 - 4 | 465 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 73, member B |
Fam73b |
5730472N09Rik, AA408683, AI790341, C9orf54, R74766 |
-10.6kb | Sense | Chr. 2 - 30364198 - 30385520 on NCBI Build GRCm38 |
| Downstream | carnitine acetyltransferase | Crat |
AW107812, CARAT |
2.9kb | Anti-sense | Chr. 2 - 30400476 - 30415748 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Dolpp1
Genomic Map