Assay Details
| Gene Symbol: Art5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111935.1 | 3 - 4 | 858 | 91 | ||
| GenBank mRNA | BC144836.1 | - | 3 - 4 | 670 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transient receptor potential cation channel, subfamily C, member 2 |
Trpc2 |
3010009O07Rik, AI115608, TRPC2a, TRPC2b, Trrp2, mTrp2, smTRPC2, trp2 |
-917.0 | Anti-sense | Chr. 7 - 102083116 - 102096864 on NCBI Build GRCm38 |
| Downstream | ADP-ribosyltransferase 1 | Art1 |
ADPRT, ARTC1, Yac-1 |
3.2kb | Anti-sense | Chr. 7 - 102101706 - 102111758 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Art5
Genomic Map