Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 2 - 3 | 265 | 99 | |
| GenBank mRNA | AK140597.1 | - | 2 - 3 | 286 | 99 | |
| AK150939.1 | - | 2 - 3 | 285 | 99 | ||
| AK156079.1 | - | 2 - 3 | 235 | 99 | ||
| AK171348.1 | - | 2 - 3 | 281 | 99 | ||
| AK178475.1 | - | 2 - 3 | 99 | |||
| BC015255.1 | - | 2 - 3 | 265 | 4221225 | 99 | |
| BC139818.1 | - | 2 - 3 | 213 | 40141268 | 99 | |
| CT010198.1 | - | 2 - 3 | 69 | 99 | ||
| D87911.1 | - | 2 - 3 | 188 | 99 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map