Assay Details
| Gene Symbol: Ackr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271607.1 | NP_001258536.1 | 3 | 907 | 63 | |
| NM_007722.4 | NP_031748.2 | 2 | 624 | 63 | ||
| GenBank mRNA | AF000236.1 | - | 2 | 578 | 63 | |
| AK031100.1 | - | 602 | 63 | |||
| BC015254.1 | - | 2 | 539 | 4242244 | 63 | |
More Information
Set Membership: |
|

Ackr3
Genomic Map