Assay Details
| Gene Symbol: Egr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289927.1 | NP_001276856.1 | 1 - 2 | 81 | 78 | |
| GenBank mRNA | BY238697.1 | - | 1 - 2 | 82 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001289927.1 transcripts.
Set Membership: |
|

Egr3
Genomic Map