Assay Details
| Gene Symbol: Peo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153796.3 | NP_722491.2 | 3 - 4 | 2168 | 93 | |
| GenBank mRNA | AK054159.1 | - | 3 - 4 | 466 | 93 | |
| AK146244.1 | - | 3 - 4 | 2062 | 93 | ||
| AY059385.1 | - | 3 - 4 | 1630 | 93 | ||
| BC034909.1 | - | 2 - 3 | 490 | 93 | ||
| BC071195.1 | - | 3 - 4 | 2168 | 5100486 | 93 | |
More Information
Set Membership: |
|

Peo1
Genomic Map